Search Thermo Fisher Scientific
Gene Symbol: XKR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289973.1 | NP_001276902.1 | 5 | 904 | ||
NM_207411.4 | NP_997294.3 | 4 | 766 | |||
XM_017013401.1 | XP_016868890.1 | 3 | 1185 | |||
GenBank mRNA | AY358489.1 | - | 4 | 637 | ||
AY534242.1 | - | 4 | 766 | |||
AY534243.1 | - | 5 | 758 |