Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABGGTA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004581.5 | NP_004572.3 | 5, 6 | 1135 | ||
NM_182836.2 | NP_878256.1 | 6, 7 | 818 | |||
GenBank mRNA | AK291347.1 | - | 6, 7 | 702 | ||
AK292613.1 | - | 5, 6 | 867 | |||
AK303247.1 | - | 4, 5 | 580 | |||
AK308715.1 | - | 6, 7 | 701 | |||
BC003093.1 | - | 6, 7 | 756 | 3537388 | ||
CR541797.1 | - | 4, 5 | 623 | |||
Y08200.1 | - | 5, 6 | 897 |