Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM72D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207418.2 | NP_997301.2 | 4 | 2182 | ||
XM_011509962.2 | XP_011508264.1 | 5 | 2217 | |||
XM_017002227.1 | XP_016857716.1 | 6 | 2602 | |||
GenBank mRNA | AB096683.1 | - | 4 | 2182 |
Gene Symbol: FAM72A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123168.2 | NP_001116640.1 | 4 | 2185 | ||
NM_001317901.1 | NP_001304830.1 | 4 | 1405 | |||
NR_134239.1 | 5 | 1465 | ||||
XM_006711526.3 | XP_006711589.1 | 5 | 2159 | |||
XM_011509965.2 | XP_011508267.1 | 5 | 2068 | |||
XM_011509967.1 | XP_011508269.1 | 4 | 2080 | |||
XM_017002233.1 | XP_016857722.1 | 5 | 2065 | |||
GenBank mRNA | BC035696.1 | - | 4 | 1752 | 5541342 | |
DQ786198.1 | - | 1 | 499 |
Gene Symbol: FAM72B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100910.1 | NP_001094380.1 | 4 | 2162 | ||
NM_001320149.1 | NP_001307078.1 | 4 | 1635 | |||
XM_011542018.1 | XP_011540320.1 | 5 | 2570 | |||
GenBank mRNA | BC046199.1 | - | 4 | 1635 | 6064384 |
Gene Symbol: FAM72C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287385.1 | NP_001274314.1 | 4 | 2200 | ||
XM_011509723.2 | XP_011508025.1 | 5 | 2253 | |||
XM_017001682.1 | XP_016857171.1 | 5 | 2450 |