Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ILKAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030768.2 | NP_110395.1 | 3, 4 | 316 | ||
XM_005246106.1 | XP_005246163.1 | 3, 4 | 309 | |||
XM_006712784.1 | XP_006712847.1 | 3, 4 | 351 | |||
XM_011511946.2 | XP_011510248.1 | 3, 4 | 313 | |||
XM_017005056.1 | XP_016860545.1 | 3, 4 | 185 | |||
XM_017005057.1 | XP_016860546.1 | 3, 4 | 699 | |||
XM_017005058.1 | XP_016860547.1 | 3, 4 | 351 | |||
GenBank mRNA | AK001043.1 | - | 3, 4 | 328 | ||
AL136850.1 | - | 3, 4 | 241 | |||
AY024365.1 | - | 3, 4 | 287 | |||
BC006576.2 | - | 3, 4 | 316 | 3604852 | ||
BC035746.1 | - | 2, 3 | 721 | |||
CR533533.1 | - | 3, 4 | 186 | |||
FJ491380.1 | - | 3, 4 | 209 | |||
FJ491381.1 | - | 3, 4 | 186 | |||
HQ658059.1 | - | 3, 4 | 316 |