Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C17orf102 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207454.2 | NP_997337.2 | 2 | 1000 | ||
GenBank mRNA | AK126768.1 | - | 2 | 1000 | ||
BC133004.1 | - | 2 | 931 | 40147026 | ||
BC136710.1 | - | 2 | 931 | 9020700 | ||
BC142651.1 | - | 2 | 984 | 40148022 |