Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM177B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324080.1 | NP_001311009.1 | 4 | 464 | ||
NM_207468.2 | NP_997351.2 | 5 | 566 | |||
NR_136691.1 | 5 | 1070 | ||||
XM_017001279.1 | XP_016856768.1 | 4 | 1008 | |||
XM_017001280.1 | XP_016856769.1 | 5 | 522 | |||
XM_017001281.1 | XP_016856770.1 | 5 | 1091 | |||
XM_017001282.1 | XP_016856771.1 | 5 | 547 | |||
XM_017001283.1 | XP_016856772.1 | 4 | 844 | |||
XM_017001284.1 | XP_016856773.1 | 5 | 437 | |||
GenBank mRNA | AK125494.1 | - | 5 | 1070 | ||
CA848039.1 | - | 4 | 416 |