Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004608.3 | NP_004599.2 | 3 | 470 | ||
XM_005255523.2 | XP_005255580.1 | 4 | 755 | |||
XM_011545926.2 | XP_011544228.1 | 4 | 918 | |||
XM_017023614.1 | XP_016879103.1 | 4 | 495 | |||
GenBank mRNA | AJ007989.1 | - | 3 | 434 | ||
AK022330.1 | - | 3 | 1104 | |||
BC026031.1 | - | 3 | 470 | 5460190 | ||
KJ534981.1 | - | 3 | 506 | |||
KJ535035.1 | - | 3 | 410 |