Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008862.3 | NP_032888.1 | 3 | 2716 | ||
GenBank mRNA | AK031123.1 | - | 1 | 1137 | ||
BC048244.1 | - | 3 | 2650 | 6308831 |