Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008867.2 | NP_032893.1 | 30 | 5156 | ||
GenBank mRNA | AK036808.1 | - | 11 | 2129 | ||
D30779.1 | - | 30 | 5109 |