Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Plxna3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008883.2 | NP_032909.2 | 25 | 4536 | ||
GenBank mRNA | AJ748653.1 | - | 7 | 1016 | ||
AK049319.1 | - | 7 | 950 | |||
AK220285.1 | - | 21 | 3160 | |||
BC093482.1 | - | 25 | 4288 | 30945372 | ||
D86950.1 | - | 25 | 4502 |