Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppm1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008910.3 | NP_032936.1 | 2 | 1103 | ||
GenBank mRNA | AF259672.1 | - | 1 | 662 | ||
AF259673.1 | - | 1 | 662 | |||
AF369981.1 | - | 1 | 672 | |||
AK076054.1 | - | 2 | 1000 | |||
AK178989.1 | - | 1 | 288 | |||
AK181077.1 | - | 1 | 289 | |||
AK194779.1 | - | 1 | 157 | |||
AK218748.1 | - | 1 | 125 | |||
BC008595.1 | - | 2 | 710 | 3581897 | ||
D28117.1 | - | 2 | 779 |