Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkar2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008924.2 | NP_032950.1 | 3, 4 | 652 | ||
GenBank mRNA | AF533977.1 | - | 3, 4 | 652 | ||
AK132728.1 | - | 3, 4 | 591 | |||
AK132824.1 | - | 3, 4 | 583 | |||
AK189797.1 | - | 1, 2 | 34 | |||
BC075623.1 | - | 3, 4 | 623 | 6854057 | ||
BC080276.1 | - | 3, 4 | 646 | 30354403 | ||
J02935.1 | - | 3, 4 | 335 |