Gene Symbol: USP9Y | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004654.3 | NP_004645.2 | 7, 8 | 1611 | ||
XM_017030078.1 | XP_016885567.1 | 6, 7 | 751 | |||
GenBank mRNA | AF000986.2 | - | 7, 8 | 1611 | ||
Y13618.1 | - | 5, 6 | 723 | |||
Y13619.1 | - | 5, 6 | 723 |