Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptger4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136079.2 | NP_001129551.1 | 3 | 2592 | ||
NM_008965.2 | NP_032991.1 | 3 | 2529 | |||
GenBank mRNA | AK034348.1 | - | 2 | 1550 | ||
AK145045.1 | - | 3 | 2529 | |||
AK156428.1 | - | 3 | 2533 | |||
BC009023.1 | - | 3 | 2087 | 3481696 | ||
BC011193.1 | - | 3 | 2577 | 4037704 | ||
D13458.1 | - | 3 | 2173 |
Set Membership: |
![]() |