Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptgs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008969.4 | NP_032995.1 | 9 | 1359 | ||
GenBank mRNA | AK046457.1 | - | 9 | 1286 | ||
AK134163.1 | - | 9 | 1245 | |||
AK138432.1 | - | 9 | 1298 | |||
AK142741.1 | - | 9 | 1286 | |||
AK159907.1 | - | 9 | 1360 | |||
AK160886.1 | - | 9 | 1278 | |||
AK163609.1 | - | 9 | 1286 | |||
AK167357.1 | - | 8 | 1237 | |||
AK170414.1 | - | 9 | 1254 | |||
AK170664.1 | - | 9 | 1286 | |||
BC005573.1 | - | 9 | 1276 | 3968101 | ||
BC023322.1 | - | 9 | 1268 | 3968136 | ||
CT010314.1 | - | 8 | 1222 | |||
M34141.1 | - | 9 | 1258 |