Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ptx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008987.3 | NP_033013.3 | 1 | 279 | ||
GenBank mRNA | BC022176.1 | - | 1 | 279 | 4012053 | |
BQ442682.1 | - | 1 | 166 | |||
CA529293.1 | - | 1 | 279 | |||
X83601.1 | - | 1 | 203 |