Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif20a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166406.1 | NP_001159878.1 | 5, 6 | 772 | ||
NM_001166407.1 | NP_001159879.1 | 5, 6 | 770 | |||
NM_009004.4 | NP_033030.1 | 5, 6 | 666 | |||
GenBank mRNA | AK051201.1 | - | 5, 6 | 584 | ||
AK051995.1 | - | 5, 6 | 621 | |||
AK053280.1 | - | 5, 6 | 584 | |||
AK083412.1 | - | 5, 6 | 626 | |||
AK083697.1 | - | 5, 6 | 586 | |||
AK084732.1 | - | 5, 6 | 608 | |||
AK142764.1 | - | 5, 6 | 596 | |||
AK144844.1 | - | 5, 6 | 584 | |||
BC060608.1 | - | 5, 6 | 772 | 5720408 | ||
Y09632.1 | - | 5, 6 | 821 |