Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ripk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009068.3 | NP_033094.3 | 11 | 2142 | ||
GenBank mRNA | AK030959.1 | - | 11 | 2373 | ||
AK050301.1 | - | 1 | 29 | |||
AK156803.1 | - | 11 | 2407 | |||
BC050905.1 | - | 11 | 2142 | 5721177 | ||
BC054542.1 | - | 11 | 2142 | 5721177 | ||
BC058162.1 | - | 11 | 2328 | 5135235 | ||
U25995.1 | - | 10 | 2057 |