Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mst1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009074.2 | NP_033100.2 | 6 | 2284 | ||
GenBank mRNA | AK053655.1 | - | 6 | 2295 | ||
BC141379.1 | - | 6 | 2095 | 9055992 | ||
X74736.1 | - | 6 | 2284 |