Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOTCH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004557.3 | NP_004548.3 | 30 | 6256 | ||
NR_134949.1 | 30 | 5825 | ||||
NR_134950.1 | 29 | 5723 | ||||
GenBank mRNA | AK131314.1 | - | 9 | 4587 | ||
BC140782.1 | - | 30 | 6211 | 9021650 | ||
BC144540.1 | - | 30 | 5786 | |||
BC144541.1 | - | 29 | 5678 | |||
D63395.1 | - | 13 | 3393 |