Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim30a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009099.2 | NP_033125.2 | 8 | 3086 | ||
GenBank mRNA | AF220014.1 | - | 8 | 2993 | ||
AF220015.1 | - | 3 | 2293 | |||
AK137592.1 | - | 1 | 1395 | |||
BC005447.1 | - | 8 | 3023 | 3493256 | ||
J03776.1 | - | 8 | 2992 |