Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009104.2 | NP_033130.1 | 4 | 465 | ||
GenBank mRNA | AK088907.1 | - | 4 | 435 | ||
AK142027.1 | - | 4 | 439 | |||
AK147111.1 | - | 4 | 432 | |||
AK161643.1 | - | 4 | 439 | |||
AK167078.1 | - | 4 | 434 | |||
AK167204.1 | - | 4 | 437 | |||
AK168117.1 | - | 4 | 436 | |||
AK168205.1 | - | 4 | 437 | |||
AK168994.1 | - | 4 | 436 | |||
BC085136.1 | - | 4 | 465 | 6402725 | ||
M14223.1 | - | 4 | 434 |