Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rxrg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159731.1 | NP_001153203.1 | 9 | 1555 | ||
NM_009107.3 | NP_033133.1 | 10 | 2011 | |||
GenBank mRNA | AK132607.1 | - | 3 | 579 | ||
BC058401.1 | - | 10 | 1993 | 5707723 | ||
S62948.1 | - | 10 | 1855 | |||
S62952.1 | - | 9 | 1511 | |||
X76656.1 | - | 1 | 220 |