Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIK3C2G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288772.1 | NP_001275701.1 | 19 | 2737 | ||
NM_001288774.1 | NP_001275703.1 | 19 | 2730 | |||
NM_004570.5 | NP_004561.3 | 18 | 2614 | |||
XM_011520696.2 | XP_011518998.1 | 19 | 2865 | |||
XM_011520697.2 | XP_011518999.1 | 19 | 2864 | |||
XM_017019470.1 | XP_016874959.1 | 19 | 2865 | |||
XM_017019471.1 | XP_016874960.1 | 19 | 2802 | |||
XM_017019472.1 | XP_016874961.1 | 20 | 3030 | |||
XM_017019473.1 | XP_016874962.1 | 19 | 3871 | |||
XM_017019474.1 | XP_016874963.1 | 18 | 2742 | |||
XM_017019475.1 | XP_016874964.1 | 19 | 2864 | |||
XM_017019476.1 | XP_016874965.1 | 17 | 2611 | |||
XM_017019477.1 | XP_016874966.1 | 18 | 2052 | |||
XM_017019479.1 | XP_016874968.1 | 6 | 776 | |||
GenBank mRNA | AJ000008.1 | - | 18 | 2639 | ||
BC130277.1 | - | 17 | 2533 | 40146308 | ||
BC144133.1 | - | 18 | 2652 | |||
BC144134.1 | - | 18 | 2656 | 9052654 | ||
BX092808.1 | - | 2 | 220 |