Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SEPT4 SIRNA Assays ›
Gene Symbol:
SEPT4
Gene Name:
septin 4
Gene Aliases:
ARTS, BRADEION, CE5B3, H5, MART, PNUTL2, SEP4, hCDCREL-2, hucep-7
Chromosome Location:
Chr.17: 58520250 - 58540818 on Build GRCh38
UniGene ID:
Hs.287518
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
15073
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SEPT4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001198713.1
NP_001185642.1
12
1493
NM_001256782.1
NP_001243711.1
13
1590
NM_001256822.1
NP_001243751.1
12
1562
NM_004574.4
NP_004565.1
12
1693
NM_080415.3
NP_536340.1
10
1732
NM_080416.3
NP_536341.1
12
1464
NR_037155.2
10
2216
NR_104196.1
11
1848
NR_104197.1
11
1587
XM_006721949.2
XP_006722012.1
13
1522
XM_006721950.3
XP_006722013.1
12
1214
XM_006721951.2
XP_006722014.1
11
3348
XM_006721952.2
XP_006722015.1
11
1167
XM_006721954.2
XP_006722017.1
12
1207
XM_006721955.2
XP_006722018.1
12
3390
XM_011524911.1
XP_011523213.1
11
1121
XM_011524912.1
XP_011523214.1
12
1879
GenBank mRNA
AB002110.1
-
10
2019
AB008753.1
-
12
1528
AF035811.1
-
12
1529
AF073312.1
-
12
1524
AF176379.1
-
10
1698
AK091065.1
-
7
1990
AK094579.1
-
11
2585
AK294094.1
-
12
1496
AK301914.1
-
13
1590
AK302146.1
-
12
1493
AK315045.1
-
11
1695
AK315396.1
-
12
1418
BC018055.1
-
11
1504
BC018056.1
-
12
1407
4811662
CR457111.1
-
12
1400
D89278.1
-
12
1400
U88829.1
-
12
1525
Back To Top
More Information
Panther Classification:
Molecular Function -
cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
cell cycle
brain development
sperm mitochondrion organization
positive regulation of protein ubiquitination
regulation of apoptotic process
positive regulation of apoptotic process
sperm capacitation
cell division
positive regulation of intrinsic apoptotic signaling pathway
GTPase activity
structural molecule activity
protein binding
GTP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-tbml7:80/10.239.166.229:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0