Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrpd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009226.4 | NP_033252.1 | 3 | 279 | ||
GenBank mRNA | AK005517.1 | - | 3 | 269 | ||
AK007674.1 | - | 3 | 280 | |||
AK011455.1 | - | 3 | 273 | |||
AK013075.1 | - | 3 | 246 | |||
AK134582.1 | - | 3 | 273 | |||
AK135979.1 | - | 3 | 269 | |||
BC024875.1 | - | 3 | 265 | 4009014 | ||
M58558.1 | - | 3 | 208 |