Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004620.3 | NP_004611.1 | 7 | 1704 | ||
NM_145803.2 | NP_665802.1 | 8 | 1797 | |||
GenBank mRNA | AK292978.1 | - | 7 | 1663 | ||
BC031052.1 | - | 8 | 1742 | 5272008 | ||
U78798.1 | - | 7 | 1637 |