Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011109.2 | NP_035239.1 | 3 | 303 | ||
GenBank mRNA | AF112983.1 | - | 3 | 259 | ||
AF124374.1 | - | 3 | 233 | |||
AF169407.1 | - | 3 | 303 | |||
AF169408.1 | - | 3 | 303 | |||
AK004232.1 | - | 3 | 264 | |||
AK018005.1 | - | 3 | 260 | |||
AK157392.1 | - | 3 | 260 | |||
AK158364.1 | - | 3 | 231 | |||
AK171797.1 | - | 3 | 263 | |||
AK172367.1 | - | 3 | 232 | |||
BC111806.1 | - | 3 | 292 | 40039182 |