Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Procr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011171.2 | NP_035301.2 | 2 | 266 | ||
GenBank mRNA | AK136541.1 | - | 2 | 126 | ||
AK143719.1 | - | 2 | 368 | |||
AK167302.1 | - | 2 | 206 | |||
BC028755.1 | - | 2 | 139 | 1054063 | ||
CT010175.1 | - | 2 | 85 | |||
L39017.1 | - | 2 | 140 | |||
X63748.2 | - | 2 | 172 |