Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PSAP SIRNA Assays ›
Gene Symbol:
Psap
Gene Name:
prosaposin
Gene Aliases:
AI037048, SGP-1
Chromosome Location:
Chr.10: 60277628 - 60302600 on Build GRCm38
UniGene ID:
Mm.277498
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
151283
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Psap
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001146120.1
NP_001139592.1
6
801
NM_001146121.1
NP_001139593.1
6
801
NM_001146122.1
NP_001139594.1
6
801
NM_001146123.1
NP_001139595.1
6
801
NM_001146124.1
NP_001139596.1
6
717
NM_011179.3
NP_035309.3
6
801
GenBank mRNA
AK088369.1
-
6
715
AK089998.1
-
6
719
AK093881.1
-
6
715
AK135637.1
-
6
715
AK145607.1
-
6
722
AK145802.1
-
6
716
AK148150.1
-
6
723
AK148152.1
-
6
742
AK148545.1
-
6
717
AK149785.1
-
6
718
AK150331.1
-
6
717
AK150645.1
-
6
715
AK151034.1
-
6
715
AK151173.1
-
6
716
AK151230.1
-
6
715
AK151234.1
-
6
717
AK151251.1
-
6
715
AK151525.1
-
6
717
AK151529.1
-
6
717
AK151686.1
-
6
751
AK151852.1
-
6
717
AK151875.1
-
6
714
AK151891.1
-
6
715
AK151907.1
-
6
717
AK151943.1
-
6
717
AK152117.1
-
6
717
AK152126.1
-
6
715
AK152134.1
-
6
717
AK152203.1
-
6
749
AK152280.1
-
6
715
AK152319.1
-
6
719
AK152355.1
-
6
717
AK152408.1
-
6
720
AK152505.1
-
6
717
AK152557.1
-
6
717
AK152605.1
-
6
717
AK152819.1
-
6
729
AK153404.1
-
6
715
AK159108.1
-
6
745
AK159164.1
-
6
715
AK159167.1
-
6
716
AK159198.1
-
6
715
AK159221.1
-
6
719
AK159244.1
-
6
715
AK159266.1
-
6
715
AK159333.1
-
6
715
AK159341.1
-
6
715
AK159384.1
-
6
715
AK159414.1
-
6
718
AK159427.1
-
6
717
AK159451.1
-
6
715
AK159457.1
-
6
729
AK159463.1
-
6
717
AK159487.1
-
6
715
AK159492.1
-
6
715
AK159510.1
-
6
715
AK159528.1
-
6
715
AK159530.1
-
6
715
AK159589.1
-
6
717
AK159604.1
-
6
715
AK159622.1
-
5
607
AK159634.1
-
6
715
AK159638.1
-
6
716
AK159657.1
-
6
715
AK159664.1
-
6
715
AK159685.1
-
6
715
AK159692.1
-
6
715
AK159709.1
-
6
715
AK159720.1
-
6
715
AK159722.1
-
6
715
AK159728.1
-
6
717
AK159840.1
-
6
715
AK159861.1
-
6
715
AK159862.1
-
6
717
AK159877.1
-
6
715
AK159910.1
-
6
715
AK159915.1
-
6
715
AK159936.1
-
6
717
AK159982.1
-
6
715
AK159994.1
-
6
715
AK160015.1
-
6
715
AK160019.1
-
6
717
AK160021.1
-
6
715
AK160024.1
-
6
715
AK160046.1
-
6
715
AK160049.1
-
6
717
AK160073.1
-
6
715
AK160108.1
-
6
715
AK160877.1
-
6
718
AK161313.1
-
6
732
AK166700.1
-
6
715
AK166707.1
-
6
714
AK166744.1
-
6
713
AK166811.1
-
6
715
AK166849.1
-
6
715
AK166879.1
-
6
717
AK166909.1
-
6
715
AK166920.1
-
6
715
AK166946.1
-
6
720
AK166959.1
-
6
715
AK166984.1
-
6
715
AK167014.1
-
6
715
AK167025.1
-
6
715
AK167073.1
-
6
715
AK167099.1
-
6
718
AK167108.1
-
6
715
AK167111.1
-
6
715
AK167147.1
-
6
715
AK167198.1
-
6
722
AK167213.1
-
6
715
AK167237.1
-
6
715
AK167243.1
-
6
716
AK167252.1
-
6
717
AK167418.1
-
6
717
AK167719.1
-
6
717
AK167902.1
-
6
785
AK170416.1
-
6
714
AK183658.1
-
15
AK189414.1
-
15
BC036266.1
-
5
581
BC141089.1
-
6
714
9055701
BC145359.1
-
6
714
9053917
S36200.1
-
6
691
S71616.1
-
6
731
U27340.1
-
6
715
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
lipid metabolic process
sphingolipid metabolic process
adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
regulation of lipid metabolic process
regulation of MAPK cascade
positive regulation of MAPK cascade
developmental growth
prostate gland growth
epithelial cell differentiation involved in prostate gland development
cellular response to organic substance
negative regulation of hydrogen peroxide-induced cell death
G-protein coupled receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-hnnbr:80/10.239.164.27:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline