Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011185.3 | NP_035315.1 | 1, 2 | 127 | ||
GenBank mRNA | AK028265.1 | - | 1, 2 | 149 | ||
AK077520.1 | - | 1, 2 | 128 | |||
AK132161.1 | - | 1, 2 | 128 | |||
AK167647.1 | - | 1, 2 | 149 | |||
BC018351.1 | - | 1, 2 | 128 | 3584538 | ||
U60824.1 | - | 1, 2 | 142 | |||
X80686.1 | - | 1, 2 | 131 |