Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ptcra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011195.2 | NP_035325.1 | 4 | 742 | ||
GenBank mRNA | BC119062.1 | - | 4 | 667 | 8733811 | |
BC120746.1 | - | 4 | 667 | 40129669 | ||
U16958.1 | - | 4 | 742 |