Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpre | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011212.3 | NP_035342.3 | 10 | 1010 | ||
GenBank mRNA | AK004602.1 | - | 10 | 1010 | ||
AK132455.1 | - | 9 | 973 | |||
AK154910.1 | - | 7 | 562 | |||
AK160947.1 | - | 7 | 559 | |||
BC053408.1 | - | 10 | 786 | |||
BC098473.1 | - | 10 | 774 | |||
D83484.1 | - | 10 | 1347 | |||
U35368.1 | - | 10 | 1010 | |||
U36758.1 | - | 7 | 559 | |||
U40280.1 | - | 9 | 750 | |||
U62387.1 | - | 10 | 912 | |||
Z23052.1 | - | 1 | 18 |