Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ranbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011240.3 | NP_035370.2 | 5 | 722 | ||
GenBank mRNA | AK040322.1 | - | 5 | 699 | ||
AK041932.1 | - | 5 | 655 | |||
AK145306.1 | - | 5 | 653 |