Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rarg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042727.2 | NP_001036192.1 | 7, 8 | 1445 | ||
NM_011244.4 | NP_035374.3 | 9, 10 | 1683 | |||
GenBank mRNA | AK148064.1 | - | 9, 10 | 1680 | ||
AK155911.1 | - | 7, 8 | 1299 | |||
BC012923.1 | - | 9, 10 | 1609 | 3154037 | ||
BC013709.1 | - | 7, 8 | 1237 | 3989967 | ||
M34475.1 | - | 7, 8 | 1221 | |||
M34476.1 | - | 9, 10 | 1440 | |||
X15848.1 | - | 9, 10 | 1503 |