Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165934.1 | NP_001159406.1 | 9 | 821 | ||
NM_011268.2 | NP_035398.2 | 9 | 821 | |||
GenBank mRNA | AF011358.1 | - | 9 | 706 | ||
AF125046.1 | - | 9 | 777 | |||
AK085443.1 | - | 9 | 747 | |||
AK138159.1 | - | 9 | 731 | |||
AK164961.1 | - | 9 | 775 | |||
AK166518.1 | - | 665 | ||||
BC129899.1 | - | 9 | 687 | 40111531 |