Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM50A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004699.3 | NP_004690.1 | 4, 5 | 555 | ||
GenBank mRNA | AD001530.1 | - | 4, 5 | 520 | ||
AK293119.1 | - | 4, 5 | 509 | |||
AK313697.1 | - | 4, 5 | 503 | |||
AL365368.1 | - | 4, 5 | 436 | |||
BC000028.2 | - | 4, 5 | 526 | 3505365 | ||
CR992165.1 | - | 4, 5 | 555 | |||
D83260.1 | - | 4, 5 | 425 |