Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011302.3 | NP_035432.3 | 6 | 1721 | ||
GenBank mRNA | AF073780.1 | - | 6 | 1584 | ||
BC046422.1 | - | 6 | 1721 | 5401012 |