Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itsn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198968.2 | NP_001185897.1 | 3 | 361 | ||
NM_001198969.2 | NP_001185898.1 | 3 | 361 | |||
NM_011365.4 | NP_035495.2 | 3 | 361 | |||
GenBank mRNA | AF132479.1 | - | 2 | 101 | ||
AF132480.1 | - | 2 | 101 | |||
AK029325.1 | - | 3 | 308 | |||
AK034367.1 | - | 3 | 362 | |||
AK042449.1 | - | 3 | 331 | |||
AK163807.1 | - | 3 | 335 | |||
BC138262.1 | - | 3 | 338 | 8861281 | ||
BC171950.1 | - | 3 | 338 | 9054236 |