Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011401.4 | NP_035531.3 | 9 | 1451 | ||
GenBank mRNA | AK028041.1 | - | 9 | 1227 | ||
AK045328.1 | - | 9 | 1227 | |||
AK147258.1 | - | 8 | 3763 | |||
AK155997.1 | - | 9 | 1210 | |||
AK164286.1 | - | 9 | 1223 | |||
BC034122.1 | - | 9 | 1209 | 4527667 | ||
BC050095.1 | - | 9 | 1205 | |||
BC058811.1 | - | 9 | 1209 | 6490844 | ||
CT010354.1 | - | 8 | 1096 |