Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stau1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109905.2 | NP_001103375.1 | 6 | 919 | ||
NM_001109906.2 | NP_001103376.1 | 6 | 937 | |||
NM_001291160.1 | NP_001278089.1 | 6 | 919 | |||
NM_011490.4 | NP_035620.1 | 6 | 913 | |||
GenBank mRNA | AF061942.1 | - | 6 | 913 | ||
AF395842.1 | - | 5 | 640 | |||
AK005002.2 | - | 6 | 860 | |||
AK139532.1 | - | 6 | 866 | |||
AK145826.1 | - | 6 | 904 | |||
AK151835.1 | - | 6 | 725 | |||
AK152269.1 | - | 6 | 731 | |||
AK152744.1 | - | 6 | 725 | |||
AK158952.1 | - | 2 | 521 | |||
AK159602.1 | - | 6 | 854 | |||
AK159781.1 | - | 6 | 889 | |||
AK160979.1 | - | 6 | 878 | |||
AK167637.1 | - | 6 | 860 | |||
AY391773.1 | - | 5 | 670 | |||
BC012959.1 | - | 6 | 883 | 4188394 | ||
BC082277.1 | - | 6 | 856 |