Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025307.1 | NP_001020478.1 | 6, 7 | 1141 | ||
NM_001286543.1 | NP_001273472.1 | 7, 8 | 1187 | |||
NM_011502.3 | NP_035632.1 | 6, 7 | 1087 | |||
NM_152220.2 | NP_689344.1 | 6, 7 | 1141 | |||
GenBank mRNA | AK038415.1 | - | 6, 7 | 648 | ||
AK133830.1 | - | 6, 7 | 664 | |||
AK170105.1 | - | 6, 7 | 550 | |||
AK171135.1 | - | 6, 7 | 684 | |||
BC024844.1 | - | 6, 7 | 549 | 5357204 | ||
BC056949.1 | - | 7, 8 | 1187 | |||
D29797.1 | - | 6, 7 | 562 | |||
D29798.1 | - | 6, 7 | 560 | |||
D29799.1 | - | 6, 7 | 575 | |||
D29800.1 | - | 7, 8 | 519 |