Gene Symbol: Sdc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011521.2 | NP_035651.1 | 4 | 365 | ||
GenBank mRNA | AK134326.1 | - | 4 | 363 | ||
AK145805.1 | - | 4 | 364 | |||
AK153445.1 | - | 4 | 365 | |||
AK159215.1 | - | 4 | 364 | |||
AK164952.1 | - | 4 | 364 | |||
BC005679.1 | - | 4 | 350 | 3154160 | ||
D89571.1 | - | 4 | 365 |