Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ptpn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163565.1 | NP_001157037.1 | 6, 7 | 1034 | ||
NM_013643.2 | NP_038671.2 | 6, 7 | 1142 | |||
GenBank mRNA | AK038416.1 | - | 6, 7 | 811 | ||
BC051975.1 | - | 6, 7 | 783 | |||
BC057339.1 | - | 5, 6 | 1304 | |||
BC079592.1 | - | 6, 7 | 1114 | 6817360 | ||
S80329.1 | - | 3, 4 | 978 | |||
U28216.1 | - | 6, 7 | 895 | |||
U28217.1 | - | 6, 7 | 1153 |