Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Sema3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013657.5 | NP_038685.3 | 18 | 4185 | ||
GenBank mRNA | AK004119.1 | - | 1 | 366 | ||
AK077231.1 | - | 7 | 3306 | |||
BC066852.1 | - | 18 | 4185 | 30093230 |