Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pstpip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013831.4 | NP_038859.3 | 843 | |||
GenBank mRNA | AK132418.1 | - | 825 | |||
AK154284.1 | - | 810 | ||||
AK154724.1 | - | 10 | 849 | |||
AK157994.1 | - | 10 | 827 | |||
BC002123.1 | - | 824 | 3494357 | |||
BC057654.1 | - | 785 | 4460817 | |||
CT010308.1 | - | 10 | 675 | |||
Y18101.1 | - | 10 | 675 |