Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc31a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290518.1 | NP_001277447.1 | 4 | 419 | ||
NM_025286.3 | NP_079562.1 | 4 | 457 | |||
GenBank mRNA | AK003269.1 | - | 4 | 392 | ||
AK007579.1 | - | 4 | 414 | |||
AK008473.1 | - | 4 | 441 | |||
AK015878.1 | - | 4 | 420 | |||
AK151612.1 | - | 4 | 387 | |||
AK152812.1 | - | 4 | 414 | |||
AK188325.1 | - | 3 | 302 | |||
BC029183.1 | - | 4 | 348 | 4502064 | ||
BC099447.1 | - | 4 | 423 | 30715212 |
Gene Symbol: Aldh9a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK177761.1 | - | 152 |