Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029391.2 | NP_083667.1 | 2 | 227 | ||
GenBank mRNA | AB232592.1 | - | 2 | 86 | ||
AF408432.1 | - | 2 | 199 | |||
AK005314.1 | - | 2 | 227 | |||
AK140513.1 | - | 2 | 160 | |||
AK170552.1 | - | 2 | 241 | |||
BC007147.1 | - | 2 | 171 | 3600193 | ||
BC096654.1 | - | 2 | 210 | 5056766 |