Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St6gal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252505.1 | NP_001239434.1 | 5 | 1147 | ||
NM_001252506.1 | NP_001239435.1 | 4 | 1014 | |||
NM_145933.4 | NP_666045.1 | 5 | 1478 | |||
GenBank mRNA | AB027197.1 | - | 2 | 317 | ||
AK084124.1 | - | 5 | 1313 | |||
BC027833.1 | - | 2 | 675 | 5321763 | ||
BC092222.1 | - | 5 | 1456 | 6844069 | ||
BC096026.1 | - | 5 | 1126 | 6833540 | ||
CF741963.1 | - | 2 | 109 | |||
D16106.1 | - | 4 | 965 |