Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136081.2 | NP_001129553.1 | 11 | 1515 | ||
NM_182990.4 | NP_892035.2 | 11 | 1644 | |||
GenBank mRNA | AK146446.1 | - | 11 | 1567 | ||
AK146585.1 | - | 11 | 1564 | |||
AK146607.1 | - | 11 | 1547 | |||
AK146691.1 | - | 11 | 1546 | |||
AK147005.1 | - | 11 | 1575 | |||
AK158552.1 | - | 11 | 1515 | |||
BC004848.1 | - | 11 | 1511 | |||
BC024835.1 | - | 8 | 1014 | |||
BC042502.1 | - | 11 | 1492 | 5005209 | ||
S50213.1 | - | 10 | 1389 |